Learn More
Zinc acetate dihydrate, 97%, specified according to the requirements of USP
CAS: 5970-45-6 | C4H6O4Zn·2H2O | 219.5 g/mol
$310.55 - $310.55
Chemical Identifiers
| CAS | 5970-45-6 |
|---|---|
| Molecular Formula | C4H6O4Zn·2H2O |
| Molecular Weight (g/mol) | 219.5 |
| InChI Key | BEAZKUGSCHFXIQ-UHFFFAOYSA-L |
| Synonym | zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn |
| PubChem CID | 2724192 |
| IUPAC Name | zinc;diacetate;dihydrate |
| SMILES | CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC451880025
|
Thermo Scientific Chemicals
451880025 |
2.5 kg | Plastic bottle |
Each for $310.55
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5970-45-6 | |
| 219.5 | |
| zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn | |
| zinc;diacetate;dihydrate |
| C4H6O4Zn·2H2O | |
| BEAZKUGSCHFXIQ-UHFFFAOYSA-L | |
| 2724192 | |
| CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2] |
Specifications
| 237.0°C | |
| 2.5 kg | |
| C4H6O4Zn·2H2O | |
| 15,10327 | |
| Solubility in water: 430g/L (20°C). Other solubilities: soluble in alcohol | |
| CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2] | |
| 219.5 | |
| 219.5 | |
| Plastic bottle | |
| 1.8400g/mL |
| 5970-45-6 | |
| 98.0 to 102% (Complexometry) | |
| (CH3CO2)2Zn·2H2O | |
| zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn | |
| BEAZKUGSCHFXIQ-UHFFFAOYSA-L | |
| zinc;diacetate;dihydrate | |
| 2724192 | |
| 97% | |
| 1.84 | |
| Zinc acetate dihydrate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Very toxic to aquatic life.
GHS P Statement
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Warning
RUO – Research Use Only