Learn More
Zinc acetate, anhydrous, 99.9+%
CAS: 557-34-6 | C4H6O4Zn | 183.468 g/mol
Supplier: Thermo Scientific Chemicals H3373418
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
ApplicationsZinc acetate anhydrous is used in the synthesis of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis. It is utilized in the ultrasonic preparation of zinc sulfide nanoparticles coated on silica particles. It is administered orally or parenterally as a nutritional supplement. It finds an application in the field of industries such as wood preservation, manufacturing other zinc salts, polymers, manufacture of ethylene acetate, as a dye mordant, and analytical reagent. It also acts as a plating inhibitor on primary water piping.
Solubility
Soluble in water, alcohol, dilute mineral acids and alkalies.
Notes
Incompatible with zinc salts, alkalies and their carbonates, oxalates, phosphates, and sulfides. Hygroscopic in nature.
Specifications
| Zinc acetate | |
| 557-34-6 | |
| C4H6O4Zn | |
| MFCD00012454 | |
| UN3077 | |
| zinc acetate, zinc diacetate, zinc ii acetate, acetic acid, zinc salt, dicarbomethoxyzinc, acetic acid, zinc ii salt, zinc di acetate, galzin, zinc acetate anhydrous, siltex cl 4 | |
| DJWUNCQRNNEAKC-UHFFFAOYSA-L | |
| zinc;diacetate | |
| 11192 | |
| 183.47 |
| anhydrous | |
| 200°C (decomposition) | |
| Zn(OOCCH3)2 | |
| 50 g | |
| Hygroscopic | |
| Soluble in water,alcohol,dilute mineral acids and alkalies. | |
| CC(=O)[O-].CC(=O)[O-].[Zn+2] | |
| 183.468 | |
| CHEBI:62984 | |
| ≥99.9% |
Chemical Identifiers
| 557-34-6 | |
| 183.468 | |
| DJWUNCQRNNEAKC-UHFFFAOYSA-L | |
| 11192 | |
| zinc;diacetate |
| C4H6O4Zn | |
| MFCD00012454 | |
| zinc acetate, zinc diacetate, zinc ii acetate, acetic acid, zinc salt, dicarbomethoxyzinc, acetic acid, zinc ii salt, zinc di acetate, galzin, zinc acetate anhydrous, siltex cl 4 | |
| CHEBI:62984 | |
| CC(=O)[O-].CC(=O)[O-].[Zn+2] |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 209-170-2
RTECSNumber : AK1500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only