missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate ACS AR Granular, Macron Fine Chemicals™

Catalog No. MK8740500
Change view
Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly
POLYSTORMOR
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK8740500 500g POLYSTORMOR
MK8740212 2.5kg Poly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK8740500 Supplier Avantor Supplier No. 874004

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.498
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

Quantity 500g
Formula Weight 219.5
Grade ACS
Packaging POLYSTORMOR
Chemical Name or Material Zinc Acetate, Dihydrate