missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, 30 Mesh, Granular, ACS, 99.8%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 125 g |
| Packaging | Poly Bottle |
| Form | Granular |
| Percent Purity | 99.8% |
| Grade | ACS |
| Chemical Name or Material | Zinc |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.