missing translation for 'onlineSavingsMsg'
Learn More

Zinc, 20 Mesh, Granular, ACS, 99.8%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor Z101525KG

 View more versions of this product

Catalog No. 18614710


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Zinc
1
Poly Bottle
Granular
99.8%
7440-66-6
2.5 kg
Approx. 20 Mesh
65.39
ACS

Chemical Identifiers

7440-66-6
65.39
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.