missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, 20 Mesh, Granular, ACS, 99.8%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor Z101512KG
Specifications
| Zinc | |
| 1 | |
| Poly Pail | |
| Granular | |
| 99.8% |
| 7440-66-6 | |
| 12 kg | |
| Approx. 20 Mesh | |
| 65.39 | |
| ACS |
Chemical Identifiers
| 7440-66-6 | |
| 65.39 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.