missing translation for 'onlineSavingsMsg'
Learn More

Zinc, 20 Mesh, Granular, ACS, 99.8%, Spectrum™ Chemical

Catalog No. 18614708
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614708 125 g Poly Bottle
18614712 500 g Poly Bottle
18614710 2.5 kg Poly Bottle
18614709 12 kg Poly Pail
18614711 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18614708 Supplier Spectrum Chemical Mfg Cor Supplier No. Z1015125GM

Chemical Identifiers

CAS 7440-66-6
Molecular Weight (g/mol) 65.39

Specifications

CAS Max % 1
Quantity 125 g
Packaging Poly Bottle
Particle Size Approx. 20 Mesh
Form Granular
Percent Purity 99.8%
Grade ACS
Chemical Name or Material Zinc Metal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.