missing translation for 'onlineSavingsMsg'
Learn More

Zinc 2-ethylhexanoate, Zn ≈ 20%, cont. 1% diethylene glycolmonomethyl ether

CAS: 136-53-8 | C16H30O4Zn | 351.792 g/mol

$100.26 - $377.78

Chemical Identifiers

CAS 136-53-8
Molecular Formula C16H30O4Zn
Molecular Weight (g/mol) 351.792
MDL Number MFCD00053316
InChI Key IFNXAMCERSVZCV-UHFFFAOYSA-L
Synonym zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate
PubChem CID 61083
IUPAC Name zinc;2-ethylhexanoate
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3956022
View Documents
Thermo Scientific Chemicals
03956022
100 g N/A
Only null left
 
AA3956036
View Documents
Thermo Scientific Chemicals
03956036
500 g N/A
Only null left
 
Description

Description

Zinc 2-ethylhexanoate is widely used in polyvinylchloride heat stabilizer, polyurethane catalyst and lubricant additive. It is an auxiliary drier used as pigments dispersing agent and prevents the paint film from webbing and frosting effects. It is involved in the preparation of highly porous dentritic zinc oxide nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

136-53-8
351.792
IFNXAMCERSVZCV-UHFFFAOYSA-L
61083
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2]
C16H30O4Zn
MFCD00053316
zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate
zinc;2-ethylhexanoate

Specifications

136-53-8
MFCD00053316
zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate
IFNXAMCERSVZCV-UHFFFAOYSA-L
zinc;2-ethylhexanoate
61083
Liquid
C16H30O4Zn
100 g
Immiscible with cold water.
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2]
351.792
351.79
Zinc 2-ethylhexanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c

H319-H361d

EINECSNumber : 205-251-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only