missing translation for 'onlineSavingsMsg'
Learn More

Zinc 2-ethylhexanoate, Zn ≈ 20%, cont. 1% diethylene glycolmonomethyl ether

Catalog No. AA3956036
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3956036 500 g
AA3956022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3956036 Supplier Thermo Scientific Chemicals Supplier No. 03956036

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 136-53-8 | C16H30O4Zn | 351.792 g/mol

Zinc 2-ethylhexanoate is widely used in polyvinylchloride heat stabilizer, polyurethane catalyst and lubricant additive. It is an auxiliary drier used as pigments dispersing agent and prevents the paint film from webbing and frosting effects. It is involved in the preparation of highly porous dentritic zinc oxide nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136-53-8
Molecular Formula C16H30O4Zn
Molecular Weight (g/mol) 351.792
MDL Number MFCD00053316
InChI Key IFNXAMCERSVZCV-UHFFFAOYSA-L
Synonym zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate
PubChem CID 61083
IUPAC Name zinc;2-ethylhexanoate
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2]

Specifications

Quantity 500 g
Solubility Information Immiscible with cold water.
Formula Weight 351.79
Physical Form Liquid
Chemical Name or Material Zinc 2-ethylhexanoate
Hazard Category H319-H361d
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
EINECSNumber 205-251-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only