missing translation for 'onlineSavingsMsg'
Learn More

Zinc 2-ethylhexanoate, ca 80% in mineral spirits (17-19% Zn)

CAS: 136-53-8 | C16H30O4Zn | 351.792 g/mol

Supplier:  Thermo Scientific Chemicals 0395610E

 View more versions of this product

Catalog No. AA395610E


Only null left

Description

Description

It is applied as a catalyst for condensation-cure silicones and as a vulcanization activator in industries. It is also employed in PVC heat stabilizer, polyurethane catalyst and lubricant additive. They are used as catalysts in paints and coatings, in polyurethanes, greases, and as wetting agents for pigments. Also used in the formulation of lubricants and greases, as hardener for epoxy resins, to make rubber and plastic products, and for refining crude oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc 2-ethylhexanoate
Colorless
C16H30O4Zn
2500 g
zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate
IFNXAMCERSVZCV-UHFFFAOYSA-L
zinc;2-ethylhexanoate
61083
Liquid
136-53-8
1.07 g/mL
MFCD00053316
UN1268
Not miscible or difficult to mix in water.
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2]
351.792
351.79

Chemical Identifiers

136-53-8
351.792
IFNXAMCERSVZCV-UHFFFAOYSA-L
61083
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2]
C16H30O4Zn
MFCD00053316
zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate
zinc;2-ethylhexanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H340-H350-H372-H304-H361-H226
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapor.

P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280i-P281-P301+P310-P303+P361+P353-P305+P351+P338-P308+P313-P331-P370+P378q-P501c

H226-H304-H319-H340-H350-H361d-H372

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.

EINECSNumber : 205-251-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only