missing translation for 'onlineSavingsMsg'
Learn More

Zinc 2-ethylhexanoate, ca 80% in mineral spirits (17-19% Zn)

Catalog No. AA3956136
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3956136 500 g
AA3956122 100 g
AA395610E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3956136 Supplier Thermo Scientific Chemicals Supplier No. 03956136

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 136-53-8 | C16H30O4Zn | 351.792 g/mol

It is applied as a catalyst for condensation-cure silicones and as a vulcanization activator in industries. It is also employed in PVC heat stabilizer, polyurethane catalyst and lubricant additive. They are used as catalysts in paints and coatings, in polyurethanes, greases, and as wetting agents for pigments. Also used in the formulation of lubricants and greases, as hardener for epoxy resins, to make rubber and plastic products, and for refining crude oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136-53-8
Molecular Formula C16H30O4Zn
Molecular Weight (g/mol) 351.792
MDL Number MFCD00053316
InChI Key IFNXAMCERSVZCV-UHFFFAOYSA-L
Synonym zinc 2-ethylhexanoate, zinc bis 2-ethylhexanoate, zinc 2-ethylhexoate, hexanoic acid, 2-ethyl-, zinc salt, hexanoic acid, 2-ethyl-, zinc salt 2:1, zinc ethylhexanoate, hexanoic acid, 2-ethyl-, zinc salt, basic, zinc 2-ethylhexanoate 100g, zinc 2+ ion bis 2-ethylhexanoate
PubChem CID 61083
IUPAC Name zinc;2-ethylhexanoate
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zn+2]

Specifications

Color Colorless
Density 1.07 g/mL
Quantity 500 g
UN Number UN1268
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 351.79
Physical Form Liquid
Chemical Name or Material Zinc 2-ethylhexanoate
Hazard Category H226-H304-H319-H340-H350-H361d-H372
Hazard Statement GHS H Statement
H340-H350-H372-H304-H361-H226
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapor.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280i-P281-P301+P310-P303+P361+P353-P305+P351+P338-P308+P313-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.
EINECSNumber 205-251-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only