missing translation for 'onlineSavingsMsg'
Learn More

Newtex™ ZetexPlus™ 200 Series Heat Resistant Gloves

Catalog No. p-7230401
Change view
Click to view available options
Length (English):
11 in.
14 in.
23 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
1139250 11 in.
1139252 14 in.
1139253 23 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1139250 Supplier Newtex™ Supplier No. 2100011
Only null left

Gloves are constructed from ZetexPlus™ Vermiculite Coated Fiberglass Fabric for protection from temperatures up to 1500°F for extended periods of time.

  • Vermiculite coating evenly dissipates heat across the surface of the glove
  • Coating significantly increases temperature rating and abrasion resistance
  • Premium wool liner
  • Double stitched and reinforced
  • Inorganic fibers will not produce hazardous outgassing

Specifications

Coating Material Vermiculite
Color Tan
Material ZetexPlus Fiberglass Fabric
Size Universal
Temperature (English) Max. 1500°F (Extended), 2000°F (Intermittent)
Temperature (Metric) Max 815°C (Extended), 1095°C (Intermittent)
For Use With (Application) Kiln/Furnace Operations, Heat Processing Facilities, and High Temperature Industrial Environments
Glove Cuff Style Open
Liner Material Wool
Series ZetexPlus™ 200
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less