missing translation for 'onlineSavingsMsg'
Learn More

Newtex™ Zetex™ 100 Series Heat Resistant Gloves

Catalog No. p-7230399
Change view
Click to view available options
Length (English):
11 in.
14 in.
23 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
1139215 11 in.
1139251 14 in.
1139259 23 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1139215 Supplier Newtex™ Supplier No. 2100005
Only null left

Gloves are constructed from Zetex™ Fiberglass Fabric for protection from temperatures up to 1000°F for extended periods of time.

  • Premium wool liner
  • Double stitched and reinforced
  • Inorganic fibers will not produce hazardous outgassing

Specifications

Coating Material Uncoated
Color White
Material Zetex Fiberglass Fabric
Size Universal
Temperature (English) Max. 1000°F (Extended), 1300°F (Intermittent)
Temperature (Metric) Max 540°C (Extended), 700°C (Intermittent)
For Use With (Application) Kiln/Furnace Operations, Heat Processing Facilities, and High Temperature Industrial Environments
Glove Cuff Style Open
Liner Material Wool
Series Zetex™ 100
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less