missing translation for 'onlineSavingsMsg'
Learn More

Zeolite ZSM-5, ammonium

Catalog No. AA4587914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4587914 25 g
AA4587922 100 g
AA4587936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4587914 Supplier Thermo Scientific Chemicals Supplier No. 04587914

Used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support. | CAS: 1318-02-1 | Al2O5Si | 162.043 g/mol

Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst supportZeolite ZSM-5 is used in hydrocarbon isomerization, alkylation of hydrocarbons. It is also used as a catalyst support. For example, copper is deposited on the zeolite and utilized for the preparation of acetaldehyde from ethanol by oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1318-02-1
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.043
MDL Number MFCD00132601
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
Synonym kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
PubChem CID 9942228
IUPAC Name dioxosilane;oxo(oxoalumanyloxy)alumane
SMILES O=[Al]O[Al]=O.O=[Si]=O

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Packaging Plastic bottle
Chemical Name or Material Zeolite ZSM-5
EINECSNumber 215-283-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only