Learn More
Zeolite Y, ammonium
CAS: 1318-02-1 | Al2O5Si | 162.043 g/mol
Supplier: Thermo Scientific Chemicals 04586722
Description
Zeolite Y is a useful metal aluminum silicate for proteomics research. It is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products. It is also used as a molecular sieve and as an adsorbent of a wide variety of cations, such as Na+, K+, Ca2+, Mg2+ and others. Zeolite is used in the development of a variety of bioreactor devices and in the determination of serum triglycerides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products
Specifications
| Zeolite Y, ammonium | |
| 1318-02-1 | |
| 100 g | |
| MFCD00132601 | |
| Insoluble in water. | |
| O=[Al]O[Al]=O.O=[Si]=O | |
| 162.043 |
| White | |
| Powder | |
| Al2O5Si | |
| kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural | |
| HNPSIPDUKPIQMN-UHFFFAOYSA-N | |
| dioxosilane;oxo(oxoalumanyloxy)alumane | |
| 9942228 |
Chemical Identifiers
| 1318-02-1 | |
| 162.043 | |
| HNPSIPDUKPIQMN-UHFFFAOYSA-N | |
| 9942228 | |
| O=[Al]O[Al]=O.O=[Si]=O |
| Al2O5Si | |
| MFCD00132601 | |
| kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural | |
| dioxosilane;oxo(oxoalumanyloxy)alumane |
Safety and Handling
EINECSNumber : 215-283-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only