missing translation for 'onlineSavingsMsg'
Learn More

Zeolite Y, ammonium

Catalog No. p-7076490
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4586914 25 g
AA4586922 100 g
AA4586936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4586914 Supplier Thermo Scientific Chemicals Supplier No. 04586914

Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products. Used in detergent manufacturing for the applications of the removal of hardness. | CAS: 1318-02-1 | Al2O5Si | 162.043 g/mol

Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1318-02-1
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.043
MDL Number MFCD00132601
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
Synonym kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
PubChem CID 9942228
IUPAC Name dioxosilane;oxo(oxoalumanyloxy)alumane
SMILES O=[Al]O[Al]=O.O=[Si]=O

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Packaging Plastic bottle
Chemical Name or Material Zeolite Y
EINECSNumber 215-283-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only