missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zeolite, type: ZSM-5
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
Specifications
Specifications
| Color | Yellow |
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| Solubility Information | Solubility in water: insoluble |
| Loss on Ignition | 10 % max. (1000°C) |
| Packaging | Plastic bottle |
| Chemical Name or Material | Zeolite |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves |
RUO – Research Use Only