missing translation for 'onlineSavingsMsg'
Learn More

Zeolite ferrierite, ammonium

Catalog No. p-7059993
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4588414 25 g
AA4588422 100 g
AA4588436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4588414 Supplier Thermo Scientific Chemicals Supplier No. 04588414
Only null left

CAS: 1318-02-1 | Al2O5Si | 162.043 g/mol

Zeolite ferrierite, ammonium is used as a commercial filter and ion-exchange bed. Further, it acts as a catalyst for the acid-catalyzed isomerization of n-butenes to isobutene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Zeolite ferrierite is used as commercial filters, ion-exchange beds, and as a catalyst for the acid-catalyzed isomerization of n-butenes to isobutene

Chemical Identifiers

CAS 1318-02-1
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.043
MDL Number MFCD00132601
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
Synonym kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, natural
PubChem CID 9942228
IUPAC Name dioxosilane;oxo(oxoalumanyloxy)alumane
SMILES O=[Al]O[Al]=O.O=[Si]=O

Specifications

Color White
Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Chemical Name or Material Zeolite ferrierite, ammonium
EINECSNumber 215-283-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only