Learn More
Zeolite beta, ammonium
Description
Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 500 g |
| Packaging | Glass bottle |
| Solubility Information | Insoluble in water. |
| Chemical Name or Material | Zeolite beta |
Safety and Handling
| EINECSNumber | 215-283-8 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only