missing translation for 'onlineSavingsMsg'
Learn More

Zenker's Fixative Solution, Ricca Chemical

Catalog No. 94101
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
94101 4 L Natural Poly Bottle
941032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 94101 Supplier Ricca Chemical Company Supplier No. 94101
Only null left

Mercuric Chloride-Potassium Dichromate-Sodium Sulfate Mixture

Specifications

Physical Form Liquid
Chemical Name or Material Zenker's Fixative Solution
Name Note Mercuric Chloride-Potassium Dichromate-Sodium Sulfate Mixture
CAS 7732-18-5, 7487-94-7, 7778-50-9, 7757-82-6
CAS Min % 90.0, 4.52, 2.26, 0.9
CAS Max % 94.0, 4.7, 2.35, 0.94
Packaging Natural Poly Bottle
Color Orange
Quantity 4 L
Grade Laboratory