Learn More
Thermo Scientific™ Zeiss 63X Objective, LD Plan-Neofluar (0.75 NA)
Catalog No. N010251
Encompass
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
N010251 1 each
1 options
Catalog No. N010251 Supplier Thermo Scientific™ Supplier No. N010251
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
Specifications
Type Objective Lens
Product Line Zeiss
Description Zeiss 63X Objective, LD Plan-Neofluar (0.75 NA)
For Use With (Equipment) ArrayScan HCA Readers
Lens Type Dry
Magnification 63X (0.75NA)
Quantity 1 each
For Research Use Only. Not for use in diagnostic procedures.