Learn More
Thermo Scientific™ Zeiss 2.5X Objective, Plan-Neofluar (0.075 NA)
Catalog No. N010250
Encompass
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
N010250 1 each
1 options
Catalog No. N010250 Supplier Thermo Scientific™ Supplier No. N010250
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
Specifications
Type Objective Lens
Product Line Zeiss
Description Zeiss 2.5X Objective, Plan-Neofluar (0.075 NA)
For Use With (Equipment) ArrayScan HCA Readers
Lens Type Dry
Magnification 2.5x (0.075NA)
Quantity 1 each
For Research Use Only. Not for use in diagnostic procedures.