missing translation for 'onlineSavingsMsg'
Learn More

Zein from Corn, TCI America™

$74.56 - $74.56

Chemical Identifiers

CAS 9010-66-6
MDL Number MFCD00062414
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
Z000125G
View Documents
TCI America
Z000125G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

9010-66-6
MFCD00062414

Specifications

9010-66-6
25 g
Zein from Corn
MFCD00062414
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'