missing translation for 'onlineSavingsMsg'
Learn More

Zein from Corn, TCI America™

Catalog No. Z000125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z000125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Z000125G Supplier TCI America Supplier No. Z000125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9010-66-6
MDL Number MFCD00062414

Specifications

Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Zein from Corn