missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zein, purified, Thermo Scientific Chemicals

$78.53 - $1020.39

Chemical Identifiers

CAS 9010-66-6
MDL Number MFCD00062414
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC179311000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
179311000
100 g Plastic bottle
Each for $78.53
 
AC179315000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
179315000
500 g Plastic bottle
Each for $204.49
 
AC179310025
SDS View Documents Promotion Details
Thermo Scientific Chemicals
179310025
2.5 kg Plastic drum
Each for $1,020.39
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9010-66-6
MFCD00062414

Specifications

9010-66-6
Plastic bottle
100 g
Solubility in water: insoluble. Other solubilities: soluble in aq. solutions ph >12,aq. ethanol,,soluble in ethanol,dmf
2% max. (800°C)
MFCD00062414
14, 10119
Zein, Purified
Safety and Handling

Safety and Handling

EINECSNumber : 232-722-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only