missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zafirlukast 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 100 mg |
| Formula Weight | 575.68 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Zafirlukast |
Safety and Handling
| RTECSNumber | FB3966200 |
| TSCA | No |