missing translation for 'onlineSavingsMsg'
Learn More

Z907 Dye Sodium Salt 85.0+%, TCI America™

Catalog No. B4432200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4432200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4432200MG Supplier TCI America Supplier No. B4432200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 871466-65-8
Molecular Formula C42H51N6NaO4RuS2
Molecular Weight (g/mol) 892.088
InChI Key OCLFVSVEFHHWRU-UHFFFAOYSA-K
Synonym Z907 Dye Sodium Salt
PubChem CID 72240744
IUPAC Name sodium;2-(4-carboxypyridin-2-yl)pyridine-4-carboxylate;4-nonyl-2-(4-nonylpyridin-2-yl)pyridine;ruthenium(2+);dithiocyanate
SMILES CCCCCCCCCC1=CC(=NC=C1)C2=NC=CC(=C2)CCCCCCCCC.C1=CN=C(C=C1C(=O)O)C2=NC=CC(=C2)C(=O)[O-].C(#N)[S-].C(#N)[S-].[Na+].[Ru+2]

Specifications

Color Black
Quantity 200 mg
Formula Weight 892.09
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(isothiocyanato)(2,2′-bipyridyl-4,4′-dicarboxylato)(4,4′-dinonyl-2,2′-bip