missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Z-Tyr-Val-Ala-Asp(OMe)-Fluoromethylketone, MP Biomedicals™

Catalog No. MP03FK01301
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP03FK01301 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP03FK01301 Supplier MP Biomedicals™ Supplier No. 03FK01301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Irreversibly inhibits activity of Caspase-1 and -4. Z-Tyr-Val-Ala-Asp(OMe)-Fluoromethylketone, MP Biomedicals peptides are potent, cell permeable, irreversible inhibitors that are used for both in vitro and in vivo applications.

Specifications

Content And Storage -20°C
Quantity 1 mg
Molecular Weight (g/mol) 630