missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-65-5 | Y | 88.91 g/mol
Supplier: Thermo Scientific Chemicals 040299DJ
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
| Yttrium rod | |
| 7440-65-5 | |
| Odorless | |
| ≈0.14g/mm | |
| (metals basis excluding Ta) | |
| MFCD00011468 | |
| VWQVUPCCIRVNHF-UHFFFAOYSA-N | |
| yttrium | |
| 23993 | |
| 99.9% |
| 6.35mm (0.25 in.) dia. | |
| 50 mm | |
| Mylar under Argon | |
| Cast Rod | |
| Y | |
| yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element | |
| [Y] | |
| 88.91 | |
| CHEBI:33331 |
Chemical Identifiers
| 7440-65-5 | |
| 88.91 | |
| VWQVUPCCIRVNHF-UHFFFAOYSA-N | |
| 23993 | |
| yttrium |
| Y | |
| MFCD00011468 | |
| yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element | |
| CHEBI:33331 | |
| [Y] |
Safety and Handling
EINECSNumber : 231-174-8
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only