missing translation for 'onlineSavingsMsg'
Learn More

Yttrium powder, -40 mesh, 99.6% (REO)

Catalog No. AA4414706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4414706 5 g
AA4414714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4414706 Supplier Thermo Scientific Chemicals Supplier No. 04414706

CAS: 7440-65-5 | Y | 88.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]

Specifications

Color Gray
Appearance Gray powder
Quantity 5 g
Mesh Size -40 Mesh
Odor Odorless
Form Powder
Assay Percent Range 99.6%
Chemical Name or Material Yttrium, Rare Earth Oxides Content
Hazard Category H228
Hazard Statement Take precautionary measures against static discharges.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation DOT Class: 4.1, Packaging Group: II
EINECSNumber 231-174-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only