missing translation for 'onlineSavingsMsg'
Learn More

Yttrium pieces, 99.9% (REO)

Catalog No. p-7122652
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle; Ampouled under argon
Under Argon
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA0061509 10 g Under Argon
AA0061518 50 g Glass bottle; Ampouled under argon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0061509 Supplier Thermo Scientific Chemicals Supplier No. 00061509
Only null left

CAS: 7440-65-5 | Y | 88.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]

Specifications

Appearance Gray pieces
Density 4.47g/cm3
Quantity 10 g
Odor Odorless
Packaging Under Argon
Form Pieces
Assay Percent Range 99.90%
Chemical Name or Material Yttrium, Rare Earth Oxides Content
EINECSNumber 231-174-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only