missing translation for 'onlineSavingsMsg'
Learn More

Yttrium Oxide, 99%+, MilliporeSigma™

Catalog No. M1124120025
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1124120025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1124120025 Supplier Technidata Supplier No. 1.12412.0025

Chemical Identifiers

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Physical Form Solid
Quantity 25 g
Assay Percent Range 99%+
Formula Weight 225.81. (molar mass)
Odor Odorless
Packaging Plastic bottle
Appearance Light gray solid
Chemical Name or Material Yttrium Oxide