Learn More
Yttrium(III) sulfide, 99.9% (REO)
CAS: 12039-19-9 | S3Y2 | 273.99 g/mol
Supplier: Thermo Scientific Chemicals 03956514
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Yttrium(III) sulfide | |
| Powder | |
| (REO) | |
| MFCD00058806 | |
| yttrium sulfide, diyttrium trisulphide, yttrium iii sulfide, yttrium sulfide y2s3, yttrium 3+ trisulfide, diyttrium 3+ ion trisulfanediide, yttrium iii sulfide trace rare earth metals basis 1g | |
| BPMRLDMEAPSVQN-UHFFFAOYSA-N | |
| diyttrium(3+) trisulfanediide | |
| 166021 | |
| 99.9% |
| 12039-19-9 | |
| 25 g | |
| S3Y2 | |
| UN3134 | |
| Soluble in water. | |
| [S--].[S--].[S--].[Y+3].[Y+3] | |
| 273.99 | |
| 273.93 | |
| Like rotten eggs |
Chemical Identifiers
| 12039-19-9 | |
| 273.99 | |
| BPMRLDMEAPSVQN-UHFFFAOYSA-N | |
| 166021 |
| S3Y2 | |
| MFCD00058806 | |
| yttrium sulfide, diyttrium trisulphide, yttrium iii sulfide, yttrium sulfide y2s3, yttrium 3+ trisulfide, diyttrium 3+ ion trisulfanediide, yttrium iii sulfide trace rare earth metals basis 1g | |
| [S--].[S--].[S--].[Y+3].[Y+3] |
Safety and Handling
GHS H Statement
H261-H315-H319-H335
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P223-P231+P232-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P335+P334-P362-P370+P378q-P501c
H261-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, TOXIC, N.O.S.
EINECSNumber : 234-887-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only