missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) sulfide, 99.9% (REO)

Catalog No. AA3956503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3956503 1 g
AA3956506 5 g
AA3956514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3956503 Supplier Thermo Scientific Chemicals Supplier No. 03956503
Only null left

CAS: 12039-19-9 | S3Y2 | 273.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12039-19-9
Molecular Formula S3Y2
Molecular Weight (g/mol) 273.99
MDL Number MFCD00058806
InChI Key BPMRLDMEAPSVQN-UHFFFAOYSA-N
Synonym yttrium sulfide, diyttrium trisulphide, yttrium iii sulfide, yttrium sulfide y2s3, yttrium 3+ trisulfide, diyttrium 3+ ion trisulfanediide, yttrium iii sulfide trace rare earth metals basis 1g
PubChem CID 166021
SMILES [S--].[S--].[S--].[Y+3].[Y+3]

Specifications

Physical Form Powder
Quantity 1 g
UN Number UN3134
Solubility Information Soluble in water.
IUPAC Name diyttrium(3+) trisulfanediide
Formula Weight 273.93
Percent Purity 99.9%
Odor Like rotten eggs
Assay (REO)
Chemical Name or Material Yttrium(III) sulfide
Hazard Category H261-H315-H319-H335
Hazard Statement GHS H Statement
H261-H315-H319-H335
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P223-P231+P232-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P335+P334-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, TOXIC, N.O.S.
EINECSNumber 234-887-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only