missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) Oxide 97.0+%, TCI America™

Catalog No. Y000725G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Y000725G 25 g
Y0007100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Y000725G Supplier TCI America Supplier No. Y000725G

Chemical Identifiers

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
Synonym Yttria
IUPAC Name Diyttrium(3+) Trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range >97.0% (T)
Merck Index 10107
Formula Weight 225.81
Percent Purity ≥97.0% (T)
Sensitivity Air Sensitive
Appearance White to Almost White Powder to Crystals
Chemical Name or Material Yttrium(III) Oxide
EINECSNumber (1)-0560
RTECSNumber ZG3850000
TSCA Yes