missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) oxide, REacton™, nanopowder, 99.995% (REO)

Catalog No. p-7059138
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4404814 25 g
AA4404822 100 g
AA4404836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4404814 Supplier Thermo Scientific Chemicals Supplier No. 04404814
Only null left

CAS: 1314-36-9 | O3Y2 | 225.81 g/mol

Yttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is presurcor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Melting Point 2410°C
Color White
Physical Form Powder
Quantity 25 g
Merck Index 14,10107
Solubility Information Insoluble in water. Soluble in alcohol and dilute acids.
Formula Weight 225.81
Percent Purity 99.995%
Grade Reacton
Odor Odorless
Sensitivity Air Sensitive
Density 5.01 g/mL
Assay (REO)
Chemical Name or Material Yttrium (III) oxide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 215-233-5
RTECSNumber ZG3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only