missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) oxide, REacton™, 99.9% (REO)

CAS: 1314-36-9 | O3Y2 | 225.81 g/mol

Supplier:  Thermo Scientific Chemicals 01118030

 View more versions of this product

Catalog No. AA1118030


Only null left
Explore available promotions

Description

Description

Yttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is presurcor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In phosphors for color TV tubes, in microwave filters, and as a stabilizer for high-temperature service materials
Specifications

Specifications

Yttrium(III) oxide
1314-36-9
Powder
99.9% (REO)
O3Y2
14,10107
RUDFQVOCFDJEEF-UHFFFAOYSA-N
diyttrium(3+) trioxidandiide
225.81
Reacton
Air Sensitive
2410°C
White
250 g
(REO)
MFCD00011473
Insoluble in water; Soluble in alcohol and dilute acids
[O--].[O--].[O--].[Y+3].[Y+3]
225.81
99.9%
Odorless
5.01 g/mL

Chemical Identifiers

1314-36-9
225.81
RUDFQVOCFDJEEF-UHFFFAOYSA-N
[O--].[O--].[O--].[Y+3].[Y+3]
O3Y2
MFCD00011473
diyttrium(3+) trioxidandiide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 215-233-5

RTECSNumber : ZG3850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only