missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) 2-ethylhexanoate, 10% w/v in n-hexane

Catalog No. AA43356AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43356AC 25 mL
AA43356AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43356AC Supplier Thermo Scientific Chemicals Supplier No. 043356AC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103470-68-4 | C24H45O6Y | 518.524 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103470-68-4
Molecular Formula C24H45O6Y
Molecular Weight (g/mol) 518.524
MDL Number MFCD00070441
InChI Key AGOMHFKGCMKLDA-UHFFFAOYSA-K
Synonym yttrium iii 2-ethylhexanoate, yttrium 2-ethylhexanoate, hexanoic acid,2-ethyl-, yttrium 3+ salt 3:1, yttrium iii 2-ethylhexanoate w/v in n-hexane, acmc-1c5ry, yttrium tri 2-ethylhexanoate, yttrium tris 2-ethylhexanoate, tris 2-ethylhexanoic acid yttrium salt, yttrium 3+ tris 2-ethylhexanoate, yttrium iii 2-ethylhexanoate y
PubChem CID 15607674
IUPAC Name 2-ethylhexanoate;yttrium(3+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Y+3]

Specifications

Quantity 25 mL
UN Number UN1208
Formula Weight 518.16
Concentration or Composition (by Analyte or Components) 10% w/v in n-hexane
Physical Form Liquid
Chemical Name or Material Yttrium (III) 2-ethylhexanoate
Hazard Category H225-H304-H315-H319-H336-H361fd-H373
Hazard Statement GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only