missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) 2,4-pentanedionate hydrate, 99.9% (REO)

Catalog No. p-7049183
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1315414 25 g
AA1315422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1315414 Supplier Thermo Scientific Chemicals Supplier No. 01315414
Only null left

CAS: 207801-29-4 | C15H21O6Y | 386.23 g/mol

Yttrium(III) 2,4-pentanedionate hydrate acts as a precursor with europium nitrate and used for the preparation of eurobium-doped yttrium orthovanadate nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 207801-29-4
Molecular Formula C15H21O6Y
Molecular Weight (g/mol) 386.23
MDL Number MFCD00013509
InChI Key CKIVBSKECCTIAG-UHFFFAOYSA-N
Synonym tris acetylacetonato yttrium n-hydrate, 3e-4-bis 2e-4-oxopent-2-en-2-yl oxy yttrio oxy pent-3-en-2-one hydrate
PubChem CID 90471001
SMILES [Y+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
IUPAC Name (E)-4-hydroxypent-3-en-2-one;yttrium;hydrate
Formula Weight 386.24 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Yttrium(III) 2,4-pentanedionate hydrate
EINECSNumber 239-607-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only