missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium pieces, sublimed, 99.9% (REO)

Catalog No. p-4408212
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4343106 5 g
AA4343114 25 g
AA4343122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4343106 Supplier Thermo Scientific Chemicals Supplier No. 04343106
Only null left

May contain up to 0.15% Calcium | CAS: 7440-64-4 | Yb | 173.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-64-4
Molecular Formula Yb
Molecular Weight (g/mol) 173.05
MDL Number MFCD00011286
InChI Key NAWDYIZEMPQZHO-UHFFFAOYSA-N
Synonym unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
PubChem CID 23992
ChEBI CHEBI:33381
IUPAC Name ytterbium
SMILES [Yb]

Specifications

Color Silver to Gray
Quantity 5 g
Assay Percent Range 99.9%
Formula Weight 173.04
Odor Odorless
Appearance Silver Gray pieces
Chemical Name or Material Ytterbium, May contain up to 0.15% Calcium
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 4.3, Packaging Group: II
EINECSNumber 231-173-2
RTECSNumber ZG1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only