Learn More
Ytterbium ingot, 99.9% (REO)
CAS: 7440-64-4 | Yb | 173.05 g/mol
$175.48 - $175.48
Chemical Identifiers
| CAS | 7440-64-4 |
|---|---|
| Molecular Formula | Yb |
| Molecular Weight (g/mol) | 173.05 |
| MDL Number | MFCD00011286 |
| InChI Key | NAWDYIZEMPQZHO-UHFFFAOYSA-N |
| Synonym | unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n |
| PubChem CID | 23992 |
| ChEBI | CHEBI:33381 |
| IUPAC Name | ytterbium |
| SMILES | [Yb] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged in air stable poly bottle
Chemical Identifiers
| 7440-64-4 | |
| 173.05 | |
| NAWDYIZEMPQZHO-UHFFFAOYSA-N | |
| 23992 | |
| ytterbium |
| Yb | |
| MFCD00011286 | |
| unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n | |
| CHEBI:33381 | |
| [Yb] |
Specifications
| 7440-64-4 | |
| Odorless | |
| Ingot | |
| MFCD00011286 | |
| NAWDYIZEMPQZHO-UHFFFAOYSA-N | |
| ytterbium | |
| 23992 | |
| 99.9% | |
| REO |
| 5 g | |
| Poly Bottle | |
| Yb | |
| unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n | |
| [Yb] | |
| 173.05 | |
| CHEBI:33381 | |
| Ytterbium ingot |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 231-173-2
TSCA : Yes
storageNote1 :
- Packaged in air stable poly bottle
Recommended Storage : Ambient temperatures
RUO – Research Use Only