Learn More
Ytterbium(III) triflate, 99.9%, (trace metal basis), anhydrous
CAS: 54761-04-5 | C3F9O9S3Yb | 620.25 g/mol
$72.73 - $72.73
Chemical Identifiers
| CAS | 54761-04-5 |
|---|---|
| Molecular Formula | C3F9O9S3Yb |
| Molecular Weight (g/mol) | 620.25 |
| InChI Key | AHZJKOKFZJYCLG-UHFFFAOYSA-K |
| Synonym | ytterbium iii trifluoromethanesulfonate, ytterbium iii triflate, ytterbium triflate, unii-4n70osw24g, ytterbium trifluoromethanesulfonate, yb otf 3, ytterbium 3+ tritriflate, trifluoromethanesulfonate; ytterbium 3+, ytterbium 3+ triflate, ytterbium trifluoromethanesulphonate |
| PubChem CID | 2733225 |
| IUPAC Name | trifluoromethanesulfonate;ytterbium(3+) |
| SMILES | C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Yb+3] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC434050010
|
Thermo Scientific Chemicals
434050010 |
1 g | Glass bottle |
Each for $72.73
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 54761-04-5 | |
| 620.25 | |
| ytterbium iii trifluoromethanesulfonate, ytterbium iii triflate, ytterbium triflate, unii-4n70osw24g, ytterbium trifluoromethanesulfonate, yb otf 3, ytterbium 3+ tritriflate, trifluoromethanesulfonate; ytterbium 3+, ytterbium 3+ triflate, ytterbium trifluoromethanesulphonate | |
| trifluoromethanesulfonate;ytterbium(3+) |
| C3F9O9S3Yb | |
| AHZJKOKFZJYCLG-UHFFFAOYSA-K | |
| 2733225 | |
| C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Yb+3] |
Specifications
| 54761-04-5 | |
| 26.8 to 29% (Yb) (Complexometry) | |
| (CF3SO3)3Yb | |
| ytterbium iii trifluoromethanesulfonate, ytterbium iii triflate, ytterbium triflate, unii-4n70osw24g, ytterbium trifluoromethanesulfonate, yb otf 3, ytterbium 3+ tritriflate, trifluoromethanesulfonate; ytterbium 3+, ytterbium 3+ triflate, ytterbium trifluoromethanesulphonate | |
| AHZJKOKFZJYCLG-UHFFFAOYSA-K | |
| trifluoromethanesulfonate;ytterbium(3+) | |
| 2733225 | |
| 99.9% | |
| Glass bottle |
| 1 g | |
| C3F9O9S3Yb | |
| 15,10304 | |
| Solubility in water: soluble | |
| C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Yb+3] | |
| 620.25 | |
| 620.25 | |
| Trace Metal Basis | |
| Ytterbium(III) triflate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
RUO – Research Use Only