missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) triflate, 99.9%, (trace metal basis), anhydrous

Catalog No. AC434050050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434050050 5 g Glass bottle
AC434050010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434050050 Supplier Thermo Scientific Chemicals Supplier No. 434050050

CAS: 54761-04-5 | C3F9O9S3Yb | 620.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54761-04-5
Molecular Formula C3F9O9S3Yb
Molecular Weight (g/mol) 620.25
InChI Key AHZJKOKFZJYCLG-UHFFFAOYSA-K
Synonym ytterbium iii trifluoromethanesulfonate, ytterbium iii triflate, ytterbium triflate, unii-4n70osw24g, ytterbium trifluoromethanesulfonate, yb otf 3, ytterbium 3+ tritriflate, trifluoromethanesulfonate; ytterbium 3+, ytterbium 3+ triflate, ytterbium trifluoromethanesulphonate
PubChem CID 2733225
IUPAC Name trifluoromethanesulfonate;ytterbium(3+)
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Yb+3]

Specifications

Quantity 5 g
Assay Percent Range 26.8 to 29% (Yb) (Complexometry)
Linear Formula (CF3SO3)3Yb
Merck Index 15,10304
Solubility Information Solubility in water: soluble
Formula Weight 620.25
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ytterbium(III) triflate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

RUO – Research Use Only