missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) chloride, 35% min w/w aq. soln., 99.9% (REO)

Catalog No. p-7058587
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42816AD 50 mL
AA42816AK 250 mL
AA42816K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42816AD Supplier Thermo Scientific Chemicals Supplier No. 042816AD

CAS: 10361-91-8 | YbCl3

Applied with NiCl2 to form a very effective catalyst for the reductive dehalogenation of aryl halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10361-91-8
Molecular Formula YbCl3
MDL Number MFCD00011288

Specifications

Physical Form Liquid
Quantity 50 mL
Solubility Information Soluble in water.
Formula Weight 279.40 (Anhydrous)
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 35% min w/w aq. soln.
Odor Odorless
Density 1.48 g/mL at 30°C
Assay (REO)
Chemical Name or Material Ytterbium (III) chloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-800-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only