missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ytterbium(III) chloride, 35% min w/w aq. soln., 99.9% (REO)
Description
Applied with NiCl2 to form a very effective catalyst for the reductive dehalogenation of aryl halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 1 L |
| Odor | Odorless |
| Density | 1.48 g/mL at 30°C |
| Solubility Information | Soluble in water. |
| Formula Weight | 279.40 (Anhydrous) |
| Percent Purity | 99.9% |
| Concentration or Composition (by Analyte or Components) | 35% min w/w aq. soln. |
| Assay | (REO) |
| Chemical Name or Material | Ytterbium (III) chloride |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 233-800-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only