missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) carbonate hydrate, REacton™, 99.9% (REO)

$649.63 - $649.63

Chemical Identifiers

MDL Number MFCD00050146
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1457418
View Documents
Thermo Scientific Chemicals
01457418
50 g
Each for $649.63
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00050146

Specifications

(decomposition)
50 g
Insoluble in water.
99.9%
Ytterbium(III) carbonate hydrate
Crystalline
MFCD00050146
526.11 (Anhydrous)
Reacton
(REO)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-585-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only