missing translation for 'onlineSavingsMsg'
Learn More

YSI™ Zobell ORP Calibration Solution

Catalog No. 1517725
Change view
Click to view available options
Filling Solution Type:
ORP Calibration Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filling Solution Type
1517725 ORP Calibration Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1517725 Supplier YSI™ Supplier No. YSI061320

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For calibrating ORP/Redox on several YSI instruments

  • A convenient temperature chart with corresponding ORP values is on the side of the container for simple calibration value inputs
  • Concentrated power makes 125mL of calibration standard
  • Plastic bottle (125mL)

Expiration Dates:
Unmixed: 2 years
Mixed: 6 months

Specifications

Type Calibration Solution
For Use With (Application) Calibrating ORP/Redox on YSI™ instruments
Filling Solution Type ORP Calibration Solution
Includes Temperature chart with corresponding ORP values is on the side of the container
Volume (Metric) Sample 119 mL
Warranty 2 Years