missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YSI™ Reagent Replacement Packs for 9300 and 9500 Photometers
$104.27 - $504.33
| Catalog Number | Mfr. No. | Type | Packaging | Range | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Packaging | Range | Price | Quantity | |||
|
15-399-12R
|
YSI™
YAP011 |
Chlorine-DPD Free | 250 Tests/Pk. | 0 to 5 mg/L |
Pack of 250 Each for $109.12
|
|
|||
|
15-399-28R
|
YSI™
YAP163 |
Nitrate | 200 Tests/Pk. | 0 to 20 mg/L |
Pack of 200 Each for $504.33
|
|
|||
|
15-399-2R
|
YSI™
YAP188 |
Alkalinity, Total | 250 Tests/Pk. | 0 to 500 mg/L |
Pack of 250 Each for $259.44
|
|
|||
|
15-399-34R
|
YSI™
YAP177 |
Phosphate LR | 200 Tests/Pk. | 0 to 4 mg/L |
Pack of 200 Each for $354.00
|
|
|||
|
15-399-29R
|
YSI™
YAP109 |
Nitrate-N | 250 Tests/Pk. | 0 to 0.5 mg/L |
Pack of 250 Each for $201.25
|
|
|||
|
15-399-33R
|
YSI™
YAP130 |
pH (Phenol Red) | 250 Tests/Pk. | 6.8 to 8.4 mg/L |
Pack of 250 Each for $104.27
|
|
|||
|
15-399-38R
|
YSI™
YAP154 |
Sulfate | 250 Tests/Pk. | 0 to 200 mg/L |
Pack of 250 Each for $145.48
|
|
|||
|
15-399-41R
|
YSI™
YAP148 |
Zinc | 250 Tests/Pk. | 0 to 4 mg/L |
Pack of 250 Each for $484.94
|
|
|||
|
15-399-4R
|
YSI™
YAP152 |
Ammonia | 250 Tests/Pk. | 0 to 1 mg/L |
Pack of 250 Each for $283.69
|
|
|||
|
15-399-7R
|
YSI™
YAP252 |
Calcium Hardness | 250 Tests/Pk. | 0 to 500 mg/L |
Pack of 250 Each for $298.23
|
|
|||
|
15-399-8R
|
YSI™
YAP268 |
Chloride | 250 Tests/Pk. | 0 to 50000 mg/L |
Pack of 250 Each for $304.94
|
|
|||
|
15-399-9R
|
YSI™
YAP031 |
Chlorine-DPD Free, Comb,Tot | 250 Tests/Pk. | 0 to 5 mg/L |
Pack of 250 Each for $180.04
|
|
|||
Specifications
| 9300 and 9500 Photometer |