missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Spectroscopy
- Spectrophotometry
- Spectrophotometer Accessories and Replacement Parts
- YSI™ Reagent Replacement Packs for 9300 and 9500 Photometers
YSI™ Reagent Replacement Packs for 9300 and 9500 Photometers
Catalog No. 153999R
Change view
Click to view available options
Type:
Alkalinity, Total
Ammonia
Calcium Hardness
Chloride
Chlorine-DPD Free
Chlorine-DPD Free, Comb,Tot
Nitrate
Nitrite-N
pH (Phenol Red)
Phosphate LR
Sulfate
Zinc
Packaging:
200 Tests/Pk.
250 Tests/Pk.
Range:
0 to 0.5 mg/L
0 to 1 mg/L
0 to 20 mg/L
0 to 200 mg/L
0 to 4 mg/L
0 to 5 mg/L
0 to 500 mg/L
0 to 50000 mg/L
6.8 to 8.4 mg/L
12 product options available for selection
| Catalog No. | Type | Packaging | Range |
|---|---|---|---|
| 153999R | Chlorine-DPD Free, Comb,Tot | 250 Tests/Pk. | 0 to 5 mg/L |
| 1539912R | Chlorine-DPD Free | 250 Tests/Pk. | 0 to 5 mg/L |
| 1539928R | Nitrate | 200 Tests/Pk. | 0 to 20 mg/L |
| 153992R | Alkalinity, Total | 250 Tests/Pk. | 0 to 500 mg/L |
| 1539934R | Phosphate LR | 200 Tests/Pk. | 0 to 4 mg/L |
| 1539929R | Nitrite-N | 250 Tests/Pk. | 0 to 0.5 mg/L |
| 1539933R | pH (Phenol Red) | 250 Tests/Pk. | 6.8 to 8.4 mg/L |
| 1539938R | Sulfate | 250 Tests/Pk. | 0 to 200 mg/L |
| 1539941R | Zinc | 250 Tests/Pk. | 0 to 4 mg/L |
| 153994R | Ammonia | 250 Tests/Pk. | 0 to 1 mg/L |
| 153997R | Calcium Hardness | 250 Tests/Pk. | 0 to 500 mg/L |
| 153998R | Chloride | 250 Tests/Pk. | 0 to 50000 mg/L |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
Specifications
Specifications
| Type | Chlorine-DPD Free, Comb,Tot |
| For Use With (Equipment) | 9300 and 9500 Photometer |
| Packaging | 250 Tests/Pk. |
| Range | 0 to 5 mg/L |