Learn More
YSI™ Probe Service Kits for YSI Dissolved Oxygen Probes
For replenishing oxygen sensor on YSI Dissolved Oxygen Probes
Supplier:  YSI™ 5776
 View more versions of this product
Catalog No. 1329932
Only null left
Description
Description
  • Membrane Kits include 150 standard, 1mil PTFE membranes or 150 high-sensitivity, 0.5mil PTFE membranes, in 10 packs of 50
  • Membrane/KCI Kits include 30 standard or high-sensitivity membranes, 1 oz. (30mL) dropper bottle with KCI crystals for adding distilled water and a probe contour tool
  • For use with probes 13-299-43 (Model 5739), 13-298-80 (Model 5750), 15-176-50 (Model 5718), 13-298-16 (Model 5300 Biological Oxygen Monitor), and 13-298-56, -57, or -62 (Model 55 Handheld Dissolved Oxygen Meters)
Specifications
Chemical Identifiers
DO
Specifications
Replenishing Oxygen Sensor on YSI™ DO Probe
30 High sensitivity membrane
30 Membranes/Pk.
High-sensitivity Membrane Kit
DO
Product Suggestions
Product Suggestions